missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ UV-Cuvette Disposable Cuvets

Fits most spectrophotometers without adapters.

Brand:  BRAND™ 759150

28.69 GBP valid until 2025-04-30
Use promo code "24155" to get your promotional price.


 View more versions of this product

Product Code. 10566581

  • £78.35 / Pack of 100
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

Description

Description

BrandTech™ BRAND™ UV-Cuvette Disposable Cuvets provide transparency from 230 to 900nm and eliminate cleaning and risk of cross-contamination. Ideal for DNA, RNA and protein analysis. Compatible with with most polar organic solvents, acids, and bases.

Advanced cuvet design includes recessed windows to protect against scratching in the optical pathway. 

  • Disposable; cleaning and cross-contamination risk are eliminated
  • Ideal for DNA, RNA, and protein analysis
  • Chemical resistant; can be used with most polar organic solvents, acids, and bases
  • Also compatible with acetone, butanone, DMF, and concentrated hydrochloric acid
  • Grouped by manufacturing mold cavity to ensure lowest variation in extinction coefficient
  • Fits most spectrophotometers without adapters 

Semi-Micro size

  • For sample volumes between 1.5 and 3.0mL
  • Available individually wrapped, DNA-, RNase-free

 

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Semi-Micro Cuvette
1.5 to 3 mL
100 Pack
230 to 900 nm
Spectrophotometers and photometers
DNA, RNA, Protein Analysis, Molecular Biology
Single-use
Plastic
10 mm
23 mm
4.5 mm
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales

Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto
BRAND™ UV-Cuvette Disposable Cuvets > Semimicro, 100/Pk.

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado