missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRAND™ BRANDplates™ pureGrade™ 96-Well Microplates
Constructed of polystyrene with nontreated surface. BRANDplates™ pureGrade™ 96-Well Microplates are noncytotoxic, and free from endotoxins, DNase, DNA, and RNase. Medium binding, nonsterile microplates are suited for homogenous assays, screening, and storage with gray alphanumeric embossing on white and black solid plates for easy well identification.
£95.20 - £755.00
Specifications
Barcode | Includes Barcode |
---|---|
Certifications/Compliance | ANSI, SLAS |
Format | Standard |
Array | 8 x 12 |
Material | Polystyrene |
Product Code | Brand | Color | Volume (Metric) Well | Well Shape | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Volume (Metric) Well | Well Shape | Price | Quantity & Availability | |||
10002213
|
BRAND™
781604 |
White | 330 μL | Round Bottom (U-well) |
£275.00
Pack of 100 |
In Stock
Log in to see stock. |
|||
10246473
|
BRAND™
781600 |
Clear | 330 μL | Round Bottom (U-well) |
£115.00
Pack of 100 |
In Stock
Log in to see stock. |
|||
10052063
|
BRAND™
781601 |
Clear | 360 μL | Conical Bottom (V-well) |
£207.00
Pack of 100 |
In Stock
Log in to see stock. |
|||
10143663
|
BRAND™
781602 |
Clear | 350 μL | Flat Bottom (F-well) |
£98.25
Pack of 100 |
In Stock
Log in to see stock. |
|||
10062063
|
BRAND™
781603 |
Clear | 350 μL | C |
£95.20
Pack of 100 |
In Stock
Log in to see stock. |
|||
10732694
|
BRAND™
781605 |
White | 350 μL | Flat Bottom (F-well) |
£257.00
Pack of 100 |
In Stock
Log in to see stock. |
|||
10448433
|
BRAND™
781607 |
Black | 330 μL | Round Bottom (U-well) |
£275.00
Pack of 100 |
In Stock
Log in to see stock. |
|||
10742694
|
BRAND™
781608 |
Black | 350 μL | Flat Bottom (F-well) |
£306.00
Pack of 100 |
Estimated Shipment: 21-08-2024
Log in to see stock. |
|||
10337723
|
BRAND™
781610 |
White | 330 μL | Flat Bottom (F-well) |
£735.00
Pack of 100 |
Estimated Shipment: 19-08-2024
Log in to see stock. |
|||
10347723
|
BRAND™
781611 |
Black, Clear Bottom | 330 μL | Flat Bottom (F-well) |
£687.00
Pack of 100 |
Estimated Shipment: 19-08-2024
Log in to see stock. |
|||
10004804
|
BRAND™
781614 |
Clear | 350 μL | Flat Bottom (F-well) |
£690.00
Pack of 50 |
In Stock
Log in to see stock. |
|||
15674007
|
BRAND™
781615 |
Black | 350 μL | Flat Bottom (F-well) |
£755.00
Pack of 50 |
In Stock
Log in to see stock. |
|||
Description
- Polystyrene surface
- Medium binding, nonsterile
- Standard plate for most applications
- Gray alphanumeric embossing on white and black solid plates for easy well identification
- Strip plates available
- Standard plates available in U-, V-, F-, C-shapes
- Transparent bottoms available in F- shape
- Available in clear, white or black
- Free from endotoxins, DNase, DNA, RNase; noncytotoxic
- Suited for homogenous assays, screening, and storage.
Specifications
Includes Barcode | |
Standard | |
Polystyrene | |
Non-sterile | |
Homogenous assays, screenings and storage |
ANSI, SLAS | |
8 x 12 | |
96 | |
Yes | |
Yes |