missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ BRANDplates™ pureGrade™ S 384-well Microplates

Non-treated sterile microplates for diverse applications

Brand:  BRAND™ 781682

239.51 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.


 View more versions of this product

Product Code. 10617903

  • £269.00 / Pack of 50
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Non-treated PS surface
  • Medium binding
  • Sterilized via beta-radiation
  • Sterile plate for most applications

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Standard
16 x 24
Polystyrene
384
Non-treated
Laboratory tasks, such as homogeneous assays, screening and storage
With Lid
Black
0.1 mL
Sterile
Flat Bottom (F-well)
50/Cs.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers