missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ Cuvet Caps

Provides tight seal, allows storage of samples to -20°C.

Brand:  BRAND™ 759242

 View more versions of this product

Product Code. 10476221

  • £16.45 / Pack of 100
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Round cuvette caps
  • Fits ultra-micro cuvettes
  • Provide tight seal
  • Allows storage of samples to -20°
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Cuvet Cap
Single-use
100 Pack
Round
Green
Polyethylene
Ultramicro UV Cuvet
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers