missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRAND™ inertGrade™ 96-Well, Non-Binding, U-Bottom Microplate
Sterile plates treated for the cultivation of non-adherent cell lines and suspension cultures.
Brand: BRAND™ 781900
378.20 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Code : 03
Additional Details : Weight : 2.71000kg
Description
- Polystyrene construction
- Sterile
- 96 wells
- U Bottom
- Transparent
- Individually wrapped, with lids
- Plates have an orange embossed alphanumeric code for easy well identification
- Surface is optimized to reduce cell adhesion, protein adsorption, enzyme and cellular activation, making this an ideal microplate surface for tumor and stem cell research
- Plates are made of virgin polystyrene and sterilized by ethylene oxide
- Manufactured to ANSI/SLAS standards 1 to 4-2004 to ensure compatibility for all processes, especially when automation is involved
Specifications
Standard | |
Individually wrapped | |
Polystyrene | |
96 | |
Non-binding | |
40 |
8 x 12 | |
Transparent | |
330 μL | |
Sterile | |
Round Bottom (U-well) |