missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRAND™ PCR Box/Rack
Holds 96-well PCR plates, tube strips, and individual 0.2mL tubes for easy handling.PCR Box/Rack is alphanumerically marked for sample identification and can be stacked (without lids). The PRC Box also holds the rack for storage.
Brand: BRAND™ 781362
36.59 GBP valid until 2025-04-30
Use promo code "24155" to get your promotional price.
This item is not returnable.
View return policy
Description
- Securely holds 96-well PCR plates, tube strips, and individual 0.2mL tubes for easy handling.
- It is alphanumerically marked for sample identification and can be stacked (without lids).
- The PCR box holds the rack for storage, and withstands temperatures from -80°C to +121°C.

Specifications
Autoclavable | |
Box/Rack | |
Polypropylene | |
98 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
BRAND™ PCR Box/Rack > PCR box/rack; Pack includes one each of red, yellow, green, purple, blue
Spot an opportunity for improvement?Share a Content Correction