Learn More
BrandTech™ PFA Sample Containers
VITLAB PFA sample containers supplied with screw cap and are suitable for trace analysis work.
Specifications
Product Type | Sample Container |
---|---|
Material | PFA |
For Use With (Application) | Sample Collection, Transport and Storage |
Description
- Suitable for sample collection, transport and storage.
- Autoclavable at 121 C
Specifications
Sample Container | |
PFA | |
Sample Collection, Transport and Storage |