Learn More
BRAND™ Polypropylene Test Tube Racks
Constructed of sturdy polypropylene.Polypropylene Test TubeRacks include alphanumeric positions for easy sample identification. Racks are stackable, non-conductive and will not float in water baths.
£49.35 - £91.59
Specifications
Autoclavable | Not Autoclavable |
---|---|
Material | Polypropylene |
Length (English) | 10.43 in. |
Width (English) | 4.96 in. |
Product Code | Brand | Color | Holds | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Holds | Price | Quantity & Availability | ||||
10603721
|
BRAND™
4340000 |
White | Tubes (13 mm) |
£74.20
Pack of 5 |
In Stock
Log in to see stock. |
||||
10014070
|
BRAND™
4340001 |
Blue | Tubes (13 mm) |
£74.20
Pack of 5 |
In Stock
Log in to see stock. |
||||
11804180
|
BRAND™
4340002 |
Red | Tubes (13 mm) |
£57.35
Pack of 5 |
In Stock
Log in to see stock. |
||||
10465421
|
BRAND™
4340011 |
Blue | Tubes (18 mm) |
£67.25
Pack of 5 |
In Stock
Log in to see stock. |
||||
10088190
|
BRAND™
4340020 |
White | Tubes (20 mm) |
£67.25
Pack of 5 |
Estimated Shipment: 14-08-2024
Log in to see stock. |
||||
11814040
|
BRAND™
4340010 |
White | Tubes (18 mm) |
£77.20
Pack of 5 |
In Stock
Log in to see stock. |
||||
10363701
|
BRAND™
4340012 |
Red | Tubes (18 mm) |
£74.20
Pack of 5 |
In Stock
Log in to see stock. |
||||
10271951
|
BRAND™
4340021 |
Blue | Tubes (20 mm) |
£57.35
Pack of 5 |
In Stock
Log in to see stock. |
||||
10150591
|
BRAND™
4340022 |
Red | Tubes (20 mm) |
£57.35
Pack of 5 |
In Stock
Log in to see stock. |
||||
10175630
|
BRAND™
4340030 |
White | Tubes (25 mm) |
£74.20
Pack of 5 |
In Stock
Log in to see stock. |
||||
10364081
|
BRAND™
4340031 |
Blue | Tubes (25 mm) |
£57.35
Pack of 5 |
In Stock
Log in to see stock. |
||||
11894150
|
BRAND™
4340040 |
White | Tubes (30 mm) |
£91.59
Pack of 5 |
In Stock
Log in to see stock. |
||||
11804160
|
BRAND™
4340041 |
Blue | Tubes (30 mm) |
£59.19
Pack of 5 |
Estimated Shipment: 14-08-2024
Log in to see stock. |
||||
11814160
|
BRAND™
4340042 |
Red | Tubes (30 mm) |
£57.35
Pack of 5 |
Estimated Shipment: 14-08-2024
Log in to see stock. |
||||
10359760
|
BRAND™
4340032 |
Red | Tubes (25 mm) |
£74.20
Pack of 5 |
In Stock
Log in to see stock. |
||||
11889550
|
BRAND™
4340060 |
White | Tubes (16 mm) |
£67.25
Pack of 5 |
In Stock
Log in to see stock. |
||||
10105830
|
BRAND™
4340062 |
Red | Tubes (16 mm) |
£57.35
Pack of 5 |
Estimated Shipment: 14-08-2024
Log in to see stock. |
||||
10760631
|
BRAND™
4340061 |
Blue | Tubes (16 mm) |
£77.20
Pack of 5 |
In Stock
Log in to see stock. |
||||
10654291
|
BRAND™
4340013 |
Yellow | Tubes (18 mm) |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
11824160
|
BRAND™
4340043 |
Yellow | Tubes (30 mm) |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
10666511
|
BRAND™
4340003 |
Yellow | Tubes (13 mm) |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
10028180
|
BRAND™
4340023 |
Yellow | Tubes (20 mm) |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
11894170
|
BRAND™
4340033 |
Yellow | Tubes (25 mm) |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
10503651
|
BRAND™
4340063 |
Yellow | Tubes (16 mm) |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
Sturdy polypropylene racks include alphanumeric positions for easy sample identification.- Racks are stackable, non-conductive and will not float in water baths.
- Suitable for use in temperatures from -20° C to 90° C.
- Supplied in three pieces for easy and permanent assembly.
- Wide spaces between positions
Specifications
Not Autoclavable | |
Polypropylene | |
10.43 in. | |
4.96 in. |