Learn More
VACUUBRAND™ Replacement 0.2 micron Biofilter for BVC Fluid Aspiration Systems
Replacement part and accessories for VACUUBRAND BVC fluid aspiration systems
Brand: VACUUBRAND™ 20638266
Code : A0
Additional Details : Weight : 0.07200kg
Description
- BVC family of fluid aspiration systems deliver compact and quiet solutions for cell culture work
- Provides flexibility to accommodate decontamination protocol whether autoclaving or disinfecting in bottle
Specifications
BVC fluid aspiration system | |
Replacement 0.2 micron Biofilter for BVC |