Learn More
BRAND™ HandyStep™ Electronic Repeating Pipetter Accessory, Battery Pack
For use with HandyStep™ Electronic Repeating Pipetter. The HandyStep™ Electronic Repeating Pipetter Accessory, Battery Pack can be easily interchanged for continuous pipetting.
Brand:  BRAND™ 705025
Code : 03
Additional Details : Weight : 0.05490kg
Product Code. 10700433
  • £40.65 / Each
In Stock
to see stock.
Description
Description
  • Replaceable nickel-metal-hydride battery pack charges in 2.5 hours (3.5 hours inside instrument)
  • Battery pack can be easily interchanged for continuous pipetting
Specifications
Specifications
NiMH
Battery Pack
HandyStep™ electronic repeating pipette battery