Learn More
BRAND™ Centrifuge Tube Mini-Cooler
Designed to protect a wide range of solutions (enzymes, DNA, RNA, cell suspensions) by helping to maintain freezer temperatures on the lab bench.
Brand: BRAND™ 114930
87.48 GBP valid until 2024-12-31
Use promo code "23264" to get your promotional price.
Code : A0
Additional Details : Weight : 0.01000kg
Description
- Manufactured from durable polycarbonate
- Filled with non-toxic gel
- Hold twelve 0.5 to 2.0mL tubes
- Provided individually
Specifications
Centrifuge tube | |
Polycarbonate | |
Red | |
0.5 to 2 mL Tubes |
Maintains 0°C temperature for 60 min. | |
Mini Cooler | |
0.5 to 2 mL |