missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRAND™ Centrifuge Tube Mini-Cooler
Designed to protect a wide range of solutions (enzymes, DNA, RNA, cell suspensions) by helping to maintain freezer temperatures on the lab bench.
Brand: BRAND™ 114935
87.48 GBP valid until 2024-12-31
Use promo code "23894" to get your promotional price.
Description
- Manufactured from durable polycarbonate
- Filled with non-toxic gel
- Hold twelve 0.5 to 2.0mL tubes
- Provided individually
Specifications
Centrifuge tube | |
Polycarbonate | |
Yellow | |
0.5 to 2 mL Tubes |
Maintains −20°C temperature for 60 min. | |
Mini Cooler | |
0.5 to 2 mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction