missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRAND™ Reagent Reservoirs
Constructed of heavy-gauge polypropylene for long-lasting durability. Reagent Reservoirs are autoclavable and suitable for all 8- and 12-channel pipetters. Non-sterile version includes lid.
£62.54 - £431.38
Specifications
Capacity (Metric) | 60 mL |
---|---|
Material | Polypropylene |
Type | Reagent Reservoir |
Autoclavable | Yes |
Product Code | Brand | Sterility | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Sterility | Packaging | Price | Quantity & Availability | ||||
10327202
|
BRAND™
703409 |
Sterile | 40 Bags of 5 |
£431.38
Pack of 200 |
Estimated Shipment: 07-08-2024
Log in to see stock. |
||||
10347392
|
BRAND™
703411 |
Sterile | Individually Wrapped |
£206.00
Pack of 100 |
In Stock
Log in to see stock. |
||||
10063970
|
BRAND™
703459 |
Non-sterile | Packs of 10 |
£62.54
Pack of 10 |
In Stock
Log in to see stock. |
||||
Description
- Heavy-gauge polypropylene
- Suitable for all 8- and 12-channel pipetters
- Autoclavable
- Non-sterile version includes lid
Specifications
60 mL | |
Polypropylene | |
Reagent Reservoir | |
Yes |