Learn More
VACUUBRAND™ Service Kits for NT Series Chemistry Pumps and Systems
Shop All Vacuubrand Products
Click to view available options
For Use With:
MD 4C NT VARIO, PC3003/4 VARIO
ME 4C NT
ME 8C NT
MZ 2C NT/VARIO, PC201 NT, PC3002 VARIO
Unit Size:
Each
Description
- Sets include appropriate diaphragms, valves and sealing rings (if applicable)
- Wrench not included.
Specifications
Specifications
| For Use With | ME 8C NT |
| Type | Service Kit |