Learn More
VACUUBRAND™ VSP3000 External Vacuum Sensor
For DCP3000 Vacuum Gauge
Brand:  VACUUBRAND™ 20636163
Product Code. 10458835
  • £456.00 / Each
Estimated Shipment: 27-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Based on thermal conductivity (Pirani) and offers a wide measurement range from atmospheric pressure down to 10-3 mbar
  • Made of plastics and ceramics with high chemical resistance, splash-water proof, also for rough operating conditions
  • Designed for use with VACUUBRAND DCP3000 Pirani fine vacuum gauge Fisher Cat 13-878-320
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Vacuubrand Vacuubus VSP3000 Pirani Fine Vacuum Gauge
Vacuum Sensor