Learn More
Branson Ultrasonics™ Ultrasonic Cleaning Bath Accessory, Covers
For use with Bransonic cleaning baths
Brand: Branson Ultrasonics™ 100-032-517
This item is not returnable.
View return policy
Description
For use with Bransonic cleaning baths
- Stainless steel lid
Spécification
Lid | |
Ultrasonic Bath, Model 2800 | |
Helps position beakers securely in bath | |
Plastic |