Learn More
R&L Enterprises™ Cork Borer
Efficiently and cleanly bores holes in cork. R&L Enterprises™ Cork Borers have a cutting tube mounted with safety under-flange in shaped handle.
Brand:  R&L Enterprises™ 3401
 View more versions of this product
Product Code. 12863952
  • £181.00 / Each
Estimated Shipment: 17-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Efficiently and cleanly bores holes in cork. R&L Enterprises™ Cork Borers have a cutting tube mounted with safety under-flange in shaped handle.
  • With rod for clearning bores
  • Manufactured from brass
  • Sizes numbered consecutively from 1 upwards in each set
Specifications
Specifications
Range: 4 to 18mm
Cork Borer
Brass