missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brass foil, alloy 260, 0.51mm (0.02 in.) thick, Thermo Scientific™
£18.00 - £34.10
Specifications
Chemical Name or Material | Brass foil, Alloy 260 |
---|---|
Name Note | 0.51mm (0.02 in.) thick |
MDL Number | MFCD00171255 |
Odor | Odorless |
Form | Foil |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15462187
|
Thermo Scientific Alfa Aesar
045181.GT |
150 x 150 mm |
£18.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15472187
|
Thermo Scientific Alfa Aesar
045181.HB |
300 x 300 mm | |||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Brass foil, Alloy 260 | |
MFCD00171255 | |
Foil |
0.51mm (0.02 in.) thick | |
Odorless |
Safety and Handling
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures