missing translation for 'onlineSavingsMsg'
Learn More

Brass foil, alloy 260, 2.0mm (0.08in) thick, Thermo Scientific Chemicals

£62.81 - £174.87

Specifications

Chemical Name or Material Brass foil, Alloy 260
Name Note 2.0mm (0.08 in.) thick
MDL Number MFCD00171255
Odor Odorless
Form Foil
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15492167
View Documents
Thermo Scientific Alfa Aesar
045137.GT
150 x 150 mm
£62.81
Each
Please to purchase this item. Need a web account? Register with us today!
15402177
View Documents
Thermo Scientific Alfa Aesar
045137.HB
300 x 300 mm
£174.87
Each
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Brass foil, Alloy 260
MFCD00171255
Foil
2.0mm (0.08 in.) thick
Odorless
Videos
Safety and Handling

Safety and Handling

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.