missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brass gauze, alloy 260, 100 mesh woven from 0.11mm (0.0045 in.) dia. wire, Thermo Scientific™
£26.30
Specifications
Chemical Name or Material | Brass gauze, Alloy 260 |
---|---|
Name Note | 100 mesh woven from 0.11mm (0.0045 in.) dia. wire |
MDL Number | MFCD00171255 |
Mesh Size | 100 Mesh |
Odor | Odorless |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15446825
|
Thermo Scientific Alfa Aesar
045208.HB |
300 x 300 mm | |||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Brass gauze, Alloy 260 | |
MFCD00171255 | |
Odorless |
100 mesh woven from 0.11mm (0.0045 in.) dia. wire | |
100 Mesh |
Safety and Handling
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures