Learn More
Cole-Parmer™ Brass Straight Compression Bulkhead Union
Offers very good chemical resistance. Ideal when working with any rigid tubing, including tubing made from fluoropolymers, other rigid plastics and metals
Brand:  Cole-Parmer™ 31412-13
 View more versions of this product
Product Code. 11825443
  • £24.25 / Each
Estimated Shipment: 24-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Very good chemical resistance.
  • Compression fittings work well with rigid tubing such as PTFE or polypropylene.
  • A compression fitting has a nut that compresses rigid tubing against fitting body for high-pressure applications.
  • Compression connector family is ideal when working with rigid tubing.
  • When selecting your fittings, consider your tubing type, material compatibility and temperature and pressure conditions.
Specifications
Specifications
Compression Bulkhead Union
Brass
-65°C
400 psi
250°C
Not Autoclavable
-85°F
1/4 in.
482°F