missing translation for 'onlineSavingsMsg'
Learn More

Masterflex™ Brass Threaded Adapter Barbed Fitting

Adapts from a NPT male thread to a hose barb for quick and easy assembly in the laboratory.

Brand:  Masterflex™ 30904-00

 View more versions of this product

Product Code. 11785788

  • £41.05 / Pack of 5
Estimated Shipment: 26-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • A crucial component in setting up a fluid handling system
  • No leaks, flow restrictions or costly downtime
  • Connections without the use of tools
  • Typically holds pressures up to 30 psi (2 bar)
  • Manufactured from brass 
  • Provided in packs of 5
Specifications

Specifications

Brass
5 Pack
Straight
Barbed Fitting
1/8 NPT(M) Thread x 3/8 in. tubing I.D.
1/8 NPT(M)
Threaded adapter
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.