Learn More
Novus Biologicals™ Breast Matched Tumor and Normal Tissue Lysate
Two matched 1mg/mL vials (100μg/vial) of human breast tissue lysate, one containing normal tissue and the other tumor tissue, for use in research. Lysates are guaranteed for 6 months from date of receipt.
Brand:  Novus Biologicals™ NBP2-47116
Product Code. 18160899
  • £490.00 / Pack of 2
Explore more special offers
Description
Description
The Breast Matched Tumor & Normal Tissue Lysate is derived from human.
Specifications
Specifications
Breast
2 Vials
Human
Store at −80°C. Avoid freeze-thaw cycles.