Learn More
BRF2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA530644
Code : PN
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: A549, HeLa, HepG2, HCT116. Predicted reactivity: Mouse (88%), Rat (90%), Bovine (91%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes one of the multiple subunits of the RNA polymerase III transcription factor complex required for transcription of genes with promoter elements upstream of the initiation site. The product of this gene, a TFIIB-like factor, is directly recruited to the TATA-box of polymerase III small nuclear RNA gene promoters through its interaction with the TATA-binding protein.Specifications
BRF2 | |
Polyclonal | |
Unconjugated | |
Brf2 | |
2700059M06Rik; 5730512K07Rik; AU018504; AU022602; B-related factor 2; BRF2; BRF-2; BRF2, RNA polymerase III transcription initiation factor 50 kDa subunit; BRF2, RNA polymerase III transcription initiation factor 50 subunit; BRF2, subunit of RNA polymerase III transcription initiation factor, BRF1-like; BRFU; FLJ11052; hBRFU; hTFIIIB50; PRO1470; RNA polymerase III transcription initiation factor BRFU; TFIIIB50; transcription factor IIB- related factor, TFIIIB50; transcription factor IIIB 50 kDa subunit | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55290 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.77 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q9HAW0 | |
Brf2 | |
Recombinant fragment corresponding to a region within amino acids 2 and 235 of Human BRF2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |