missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brij™ C20, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
500 g
2.5 kg
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Color | White |
Acid Value | 1mg KOH/g max. |
Physical Form | Pellets |
Melting Point | 35.4°C to 39.6°C |
Linear Formula | C16H33(OCH2CH2)20OH |
Quantity | 5 g |
Merck Index | 10, 7449 |
Solubility Information | Solubility in water: soluble. Other solubilities: soluble in lower alcohols, insoluble in propylene glycol, coal-tar hydrocarbo, n, cottonseed and mineral oil |
Flash Point | >149°C |
IUPAC Name | 2-[2-[2-[2-[2-[2-[2-[2-[2-[2-[2-[2-[2-[2-[2-[2-[2-[2-[2-(2-hexadecoxyethoxy)ethoxy]ethoxy]ethoxy]ethoxy]ethoxy]ethoxy]ethoxy]ethoxy]ethoxy]ethoxy]ethoxy]ethoxy]ethoxy]ethoxy]ethoxy]ethoxy]ethoxy]ethoxy]ethanol |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'