missing translation for 'onlineSavingsMsg'
Learn More

Brij∣r L23, Electrophoresis Reagent

Product Code. 15457329
Click to view available options
Quantity:
250 g
1000 g
Unit Size:
1000g
250g
This item is not returnable. View return policy

Product Code. 15457329

Brand: Thermo Scientific Alfa Aesar J66096.0B

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 9002-92-0

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Brij™ L23
Color White
Name Note Electrophoresis Reagent
CAS 9002-92-0
Physical Form Waxy Solid
Melting Point ∼35°C to 40°C
Quantity 1000 g
MDL Number MFCD00080891
Synonym Brij™ 35; Tricosaethylene glycol mono-n-dodecyl ether
Solubility Information Soluble in water
Formula Weight 1199.57
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.