Learn More
Brij∣r L23, Thermo Scientific Chemicals
£20.20 - £203.01
Chemical Identifiers
CAS | 9002-92-0 |
---|---|
Molecular Formula | C14H30O2 |
Molecular Weight (g/mol) | 230.39 |
MDL Number | MFCD00043063 |
InChI Key | SFNALCNOMXIBKG-UHFFFAOYSA-N |
IUPAC Name | 2-(dodecyloxy)ethan-1-ol |
SMILES | CCCCCCCCCCCCOCCO |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11432478
|
Thermo Scientific Alfa Aesar
A15809.30 |
250 g |
£20.20
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11422478
|
Thermo Scientific Alfa Aesar
A15809.0B |
1000 g |
£48.24
1000g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11442478
|
Thermo Scientific Alfa Aesar
A15809.0I |
5000 g |
£203.01
5000g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Brij™ L23 is use in Stein-Moore chromatography. Used in the schlerotherapy treatments, in those suffering from blood vessel malformations and mutations. It acts as a surfactant in Micellar electrokinetic chromatography (MEKC).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
SolubilitySoluble in water (40 mg/ml) at 20°C methanol, chloroform and ethanol.
Notes
Store at room temperature. Store in cool dry place in tightly closed container. With good ventilation. Store away from oxidizing agent. Used as a wetting agent, a detergent, or an emulsifier. Brij™L23 is a detergent used for measuring the concentration of carbon black in lung tissues.
Chemical Identifiers
9002-92-0 | |
230.39 | |
SFNALCNOMXIBKG-UHFFFAOYSA-N | |
CCCCCCCCCCCCOCCO |
C14H30O2 | |
MFCD00043063 | |
2-(dodecyloxy)ethan-1-ol |
Specifications
9002-92-0 | |
C14H30O2 | |
MFCD00043063 | |
SFNALCNOMXIBKG-UHFFFAOYSA-N | |
2-(dodecyloxy)ethan-1-ol | |
1199.57 |
∽35°C to 40°C | |
CH3(CH2)10CH2(OCH2CH2)23OH | |
Soluble in water (40mg/ml) at 20°C methanol,chloroform and ethanol. | |
CCCCCCCCCCCCOCCO | |
230.39 | |
Brij 35 |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P362-P301+P312-P312-P405-P403+P233-P501a
Warning
missing translation for 'einecsNumber' : 500-002-6
missing translation for 'rtecsNumber' : JR5990000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Brij L23
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P405-Store locked up.
- P501-Dispose of contents/container to in accordance with local/regional/national/international regulations..
RUO – Research Use Only