missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Brilliance™ Candida Agar

Obtain fast, accurate results with a selective differential chromogenic medium that isolates and identifies clinically relevant Candida spp., including C. albicans.

Brand:  Thermo Scientific™ PO1034A

23.36 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.


 View more versions of this product

Product Code. 12987188

  • £50.25 / Pack of 10

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description
Show More
Specifications

Specifications

Off white, opaque
Brilliance Candida Agar
United Kingdom
19.5g ± 1.0g
10/Pk.
2°C to 10°C
90mm Monoplate
Agar
Flow wrapped - single layer
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number4499850 Certificate TypeCertificate of Analysis Date24/03/2025 Catalog NumberPO1034A
Lot Number4497624 Certificate TypeCertificate of Analysis Date17/03/2025 Catalog NumberPO1034A
Lot Number4495874 Certificate TypeCertificate of Analysis Date07/03/2025 Catalog NumberPO1034A
Lot Number4496119 Certificate TypeCertificate of Analysis Date04/03/2025 Catalog NumberPO1034A
Lot Number4492334 Certificate TypeCertificate of Analysis Date21/02/2025 Catalog NumberPO1034A
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Brilliance™ Candida Agar >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.