missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Brilliance™ GBS Agar

Product Code. 15138824
Click to view available options
Description:
Brilliance GBS Agar
Quantity:
10/Pk.
Manufacturing Origin:
Germany
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15138824

Brand: Thermo Scientific™ PO5320A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Screen for the presence of group B streptococci in clinical samples with the only chromogenic medium that doesn't require pre-enrichment or re-incubation of negatives.

Eliminate the need for pre-enrichment or re-incubation using Thermo Scientific™ Brilliance™ GBS Agar, the first chromogenic media for Group B Streptococci which doesn’t require extra steps. Simply streak the sample onto the plate, incubate and read your result after 24 hours. Brilliance™GBS Agar saves up to 72 hours compared to traditional media and up to 48 hours compared to other chromogenic media.

Common methods for Group B Streptococci screening use traditional media which require up to 72 hours incubation. Furthermore, additional consumables are required to generate the anaerobic atmosphere for the growth of GBS. Commercially available chromogenic media allow aerobic incubation and make the additional consumables unnecessary; however, most still require pre-enrichment and re-incubation of negatives which prolong the GBS diagnostic process. Not Brilliance GBS Agar.

This selective medium for screening for the presence of group B streptococci carried in vaginal and vaginal-rectal clinical samples allow for presumptive identification and accurate differentiation of GBS with a second chromogen. Non-GBS grow as blue or purple colonies on Brilliance GBS Agar. The formulation also incorporates Inhibigen technology, a targeted inhibiton of enterococci and group D streptococci, ensuring a high level of sensitivity and specificity.

This product may not be available in all countries or territories. Please consult your local representative for availability.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
TRUSTED_SUSTAINABILITY

Specifications

Color Oyster white, transparent
Content And Storage 2° to 12°C, in the dark
Description Brilliance GBS Agar
Format 90mm Monoplate
Manufacturing Origin Germany
Product Type Agar
Fill Weight 17g ± 5%
Packaging Type Flow wrapped - single layer
Quantity 10/Pk.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.