missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Brilliance™ MRSA 2 Agar

Product Code. p-7000001
Click to view available options
Quantity:
10/Pk.
Format:
90mm Monoplate
Media Type:
Brilliance MRSA 2 Agar
Manufacturing Origin:
United Kingdom
Unit Size:
Pack of 10
This item is not returnable. View return policy

Product Code. 12947208

Brand: Thermo Scientific™ PO1210A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Obtain fast and accurate results with a selective chromogenic medium for screening clinical samples for the presence of Methicillin Resistant Staphylococcus aureus (MRSA).

Obtain results after 18 hours with no need for re-incubation of negatives using Thermo Scientific™ Brilliance™ MRSA 2 Agar. Rapidly eliminate negative results and target colonized patients for isolation and appropriate treatment as early as possible.

Brilliance MRSA 2 Agar has been enhanced over that of the original formulation in two ways. New inhibitory components in the medium inhibit the growth of more non-target organisms. In addition, those organisms that do grow are more easily distinguished from distinctive blue MRSA colonies, through inclusion of a novel pink counter-stain, further improving ease of interpretation.

Brilliance MRSA 2 Agar has been refined to give larger MRSA colonies a more consistent color than the previous formulation. The improved recovery of MRSA, together with the increased selectivity, makes the new Brilliance MRSA 2 Agar an invaluable and efficient tool in the implementation of infection control.
  • Brilliance MRSA 2 Agar can be inoculated from a screening swab taken from hospital patient or staff, from an isolated colony or from a liquid suspension.
  • MRSA grows as blue colonies which are very easy to read against the light colored, opaque background.
  • Plates are incubated at 37ºC, and provide high sensitivity and specificity, with results available in just 18 hours. This allows a rapid response, so enabling the patient to receive the most appropriate treatment as early as possible, reducing the risk of onward transmission of infection.
  • Accuracy minimizes costs by helping to ensure that only those in need receive what can be costly treatment.
Our products are manufactured and distributed across the world through a vast network of company-owned facilities to ensure you receive the freshest, highest performing medium possible. For fastest delivery we recommend you select the product part number with a manufacturing origin closest to you.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
TRUSTED_SUSTAINABILITY

Specifications

Color Pale buff, opaque
Content And Storage 2°C to 10°C
Format 90mm Monoplate
Manufacturing Origin United Kingdom
Media Type Brilliance MRSA 2 Agar
Product Type Agar
Fill Weight 19.5g ± 1.0g
Packaging Type Flow Wrapped - Single Layer
Quantity 10/Pk.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.