Learn More
Thermo Scientific™ Brilliance™ Salmonella
Identify Salmonella spp. in food with this chromogenic medium that when used with One Broth Salmonella can shave two days off results time compared to other methods.
Brand: Thermo Scientific™ PO5098A
28.63 GBP valid until 2025-07-13
Use promo code "24135" to get your promotional price.
Description
Quickly identify Salmonella spp. with Thermo Scientific™ Brilliance™ Salmonella, containing proprietary Inhibigen technology. Use in conjunction with One Broth Salmonella to streamline your workflow and achieve faster results compared to traditional methods.
Brilliance Salmonella Agar provides a highly selective and easy-to-read medium for the presumptive identification of Salmonella from food and environmental samples.The Inhibigen contained in this medium specifically targets Escherichia coli with additional compounds added to suppress the growth of non-target organisms. Differentiation of Salmonella is achieved through the inclusion of two chromogens that target specific enzymes: caprylate esterase and ß-glucosidase. The color produced depends on which enzymes the organism possesses.
Differentiation is easy with puple Salmonella colonies and blue colonies from non-target organisms.
For faster results during food testing, use with One Broth Salmonella (CM1091B) to obtain results within two days.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.

Specifications
Cream to pink tinged, opaque | |
Brilliance Salmonella | |
Germany | |
17g ± 5% | |
10/Pk. |
2° to 12°C, in the dark | |
90mm Monoplate | |
Agar | |
Flow Wrapped - Single Layer |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
6258135 | Certificate of Analysis | 13/04/2025 | PO5098A |
6258136 | Certificate of Analysis | 13/04/2025 | PO5098A |
6258137 | Certificate of Analysis | 13/04/2025 | PO5098A |
6255059 | Certificate of Analysis | 03/04/2025 | PO5098A |
6255058 | Certificate of Analysis | 03/04/2025 | PO5098A |
Special Offers
-
Multiple Manufacturers
Elevate Your Beverage Testing with Precision Solutions
Get amazing savings on products for your analysesPromo Code: 24135Expires: 2025-07-13
Days
83Hours
12Minutes
13Seconds
34New Offer Save Up to 85%
Your input is important to us. Please complete this form to provide feedback related to the content on this product.