missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Brilliance™ Staph 24 Agar

Product Code. 12678767
Click to view available options
Description:
Brilliance Staph 24 Agar
Quantity:
10/Pk.
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 12678767

Brand: Thermo Scientific™ PO1186A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Isolate and enumerate coagulase-positive staphylococci in food or clinical samples in just 24 hours with selective, chromogenic Thermo Scientific™ Brilliance™ Staph 24 Agar.

Isolate and enumerate coagulase-positive staphylococci in food or clinical samples in just 24 hours with selective, chromogenic Thermo Scientific™ Brilliance™ Staph 24 Agar. Coagulase-positive staphylococci (CPS) grow as dark blue colonies on a clear background, making results much easier to read than traditional Baird-Parker Agar formulations.

Isolate and enumerate coagulase-positive staphylococci from food or clinical samples in just 24 hours with this selective, chromogenic medium to aid rapid food safety assessment or to facilitate faster patient treatment.

  • Results in half the time: 24 hours, compared to the 48 hours required for Baird-Parker Egg Yolk Tellurite Agar (BP-EYT)
  • Fewer confirmatory tests: selective agents have been carefully designed to inhibit the growth of Gram-negative flora and non-target Gram-positive organisms
  • Easy-to-read test: the chromogen is specifically activated by CPS which colors positive colonies dark blue, while coagulase-negative staphylococci are inhibited or remain colorless
  • Ready-to-use convenience of prepared media

Foods that require extensive handling and preparation or that are kept at slightly elevated temperatures after preparation are commonly involved in staphylococcal food poisoning.

Most species of staphylococci can produce some form of enterotoxin, the causative agent of staphylococcal enteritis. The production of these enterotoxins strongly correlates with the production of coagulase and/or thermonuclease1, both of which are used as markers of pathogenicity. When coagulase-positive staphylococci are prevalent in low numbers, as they are in many foods, they pose a limited risk of infection. However CPS numbers in excess of 1x105 cells/g are considered sufficient to produce enough enterotoxin to induce gastroenteritis.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 10°C
Description Brilliance Staph 24 Agar
Format 90mm Monoplate
Product Type Agar
Fill Weight 19.5g ± 1g
Packaging Type Wrapped in single cellulose-based film wrap
Quantity 10/Pk.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.