missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Brilliance™ UTI Clarity
Click to view available options
Quantity:
10/Pk.
Format:
90mm Monoplate
Media Type:
Brilliance UTI Clarity
Manufacturing Origin:
United Kingdom
Unit Size:
Pack of 10
Description
Enumerate, identify and isolate common organisms associated with urinary tract infections within 18 to 24 hours using Thermo Scientific™ Brilliance™ UTI Clarity with no further confirmation for E.coli. Organisms are differentiated through the utilization of one or more chromogens and/or tryptophan. Interpretation is aided by the inclusion of an transparent background, providing a distinct contrast against the seven possible colors of colony growth.
The Brilliance UTI Clarity formulation contains two chromogenic substrates which are cleaved by the ß-galactosidase and ß-glucosidase enzymes produced by E. coli, Enterococcus species and coliforms. These specific enzyme reactions cleave the chromogens giving a range of diagnostic colors.
The ß-galactosidase activity of E. coli and S. saprophyticus results in pink/red colonies and the ß-glucosidase activity of enterococci produces blue/turquoise colonies; the activity of both enzymes on coliforms gives dark blue/purple colonies. Tryptophan deaminase activity produces a brown halo around colonies of Proteus, Morganella and Providencia species. Most other organisms exhibit their natural pigmentation.
The specificity of Brilliance UTI clarity minimises confirmatory testing, with same day results for E.coli confirmation being achieved using a rapid indole test (DMAC) (MB1448A).
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
The ß-galactosidase activity of E. coli and S. saprophyticus results in pink/red colonies and the ß-glucosidase activity of enterococci produces blue/turquoise colonies; the activity of both enzymes on coliforms gives dark blue/purple colonies. Tryptophan deaminase activity produces a brown halo around colonies of Proteus, Morganella and Providencia species. Most other organisms exhibit their natural pigmentation.
The specificity of Brilliance UTI clarity minimises confirmatory testing, with same day results for E.coli confirmation being achieved using a rapid indole test (DMAC) (MB1448A).
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifications
Specifications
| Color | Straw, clear |
| Content And Storage | 2°C to 10°C |
| Format | 90mm Monoplate |
| Manufacturing Origin | United Kingdom |
| Media Type | Brilliance UTI Clarity |
| Product Type | Agar |
| Fill Weight | 19.5g ± 1.0g |
| Packaging Type | Flow Wrapped - Single Layer |
| Quantity | 10/Pk. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction