missing translation for 'onlineSavingsMsg'
Learn More

Coomassie Brilliant Blue G-250, Fisher BioReagents

Product Code. 11824422
Click to view available options
Quantity:
25 g
50 g
Packaging:
Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy
Brilliant Blue G-250 is a general protein stain.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6104-58-1
Molecular Formula C47H48N3NaO7S2
Molecular Weight (g/mol) 854.025
InChI Key RWVGQQGBQSJDQV-UHFFFAOYSA-M
Synonym Coomassie™ Blue G-250
PubChem CID 6328534
IUPAC Name sodium;3-[[4-[(Z)-[4-(4-ethoxyanilino)phenyl]-[4-[ethyl-[(3-sulfonatophenyl)methyl]azaniumylidene]-2-methylcyclohexa-2,5-dien-1-ylidene]methyl]-N-ethyl-3-methylanilino]methyl]benzenesulfonate
SMILES CCN(CC1=CC(=CC=C1)S(=O)(=O)[O-])C2=CC(=C(C=C2)C(=C3C=CC(=[N+](CC)CC4=CC(=CC=C4)S(=O)(=O)[O-])C=C3C)C5=CC=C(C=C5)NC6=CC=C(C=C6)OCC)C.[Na+]

Specifications

Extinction Coefficient 600g(-1) cm(-1) min.
Identification Pass Test
Lambda Max 608 to 612nm
Formula Weight 854.02
Loss on Drying 5% max.
Packaging Glass Bottle
Color Index No. 42655
Color Blue
Quantity 25 g
Physical Form Solid
Chemical Name or Material Brilliant Blue G-250
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'