missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Brilliant Blue G, pure
£20.20 - £556.77
Chemical Identifiers
CAS | 6104-58-1 |
---|---|
Molecular Formula | C47H48N3NaO7S2 |
Molecular Weight (g/mol) | 854.025 |
MDL Number | MFCD00078482 |
InChI Key | RWVGQQGBQSJDQV-UHFFFAOYSA-M |
Synonym | Acid Blue 90, Brilliant Blue G 250, C.I. 42655 |
PubChem CID | 6328534 |
IUPAC Name | sodium;3-[[4-[(Z)-[4-(4-ethoxyanilino)phenyl]-[4-[ethyl-[(3-sulfonatophenyl)methyl]azaniumylidene]-2-methylcyclohexa-2,5-dien-1-ylidene]methyl]-N-ethyl-3-methylanilino]methyl]benzenesulfonate |
SMILES | CCN(CC1=CC(=CC=C1)S(=O)(=O)[O-])C2=CC(=C(C=C2)C(=C3C=CC(=[N+](CC)CC4=CC(=CC=C4)S(=O)(=O)[O-])C=C3C)C5=CC=C(C=C5)NC6=CC=C(C=C6)OCC)C.[Na+] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10266390
|
Thermo Scientific Chemicals
191480050 |
5 g | Glass bottle |
£20.20
5g |
In Stock
Log in to see stock. |
||||
10358090
|
Thermo Scientific Chemicals
191480250 |
25 g | Glass bottle |
£61.61
25g |
In Stock
Log in to see stock. |
||||
15710829
|
Thermo Scientific Chemicals
191481000 |
100 g | Glass bottle |
£164.82
100g |
In Stock
Log in to see stock. |
||||
10398933
|
Thermo Scientific Chemicals
191485000 |
500 g | Glass bottle |
£556.77
500g |
In Stock
Log in to see stock. |
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
6104-58-1 | |
854.025 | |
RWVGQQGBQSJDQV-UHFFFAOYSA-M | |
6328534 | |
CCN(CC1=CC(=CC=C1)S(=O)(=O)[O-])C2=CC(=C(C=C2)C(=C3C=CC(=[N+](CC)CC4=CC(=CC=C4)S(=O)(=O)[O-])C=C3C)C5=CC=C(C=C5)NC6=CC=C(C=C6)OCC)C.[Na+] |
C47H48N3NaO7S2 | |
MFCD00078482 | |
Acid Blue 90, Brilliant Blue G 250, C.I. 42655 | |
sodium;3-[[4-[(Z)-[4-(4-ethoxyanilino)phenyl]-[4-[ethyl-[(3-sulfonatophenyl)methyl]azaniumylidene]-2-methylcyclohexa-2,5-dien-1-ylidene]methyl]-N-ethyl-3-methylanilino]methyl]benzenesulfonate |
Specifications
425 (at lambda min.) | |
6104-58-1 | |
60% min. | |
MFCD00078482 | |
Acid Blue 90, Brilliant Blue G 250, C.I. 42655 | |
RWVGQQGBQSJDQV-UHFFFAOYSA-M | |
CCN(CC1=CC(=CC=C1)S(=O)(=O)[O-])C2=CC(=C(C=C2)C(=C3C=CC(=[N+](CC)CC4=CC(=CC=C4)S(=O)(=O)[O-])C=C3C)C5=CC=C(C=C5)NC6=CC=C(C=C6)OCC)C.[Na+] | |
854.025 | |
608 to 618nm (in 50:50 IMS:H2O) | |
10% max. (105°C) | |
Crystalline Powder |
>100.0°C | |
0.95 to 1.15 | |
C47H48N3NaO7S2 | |
14, II, 450 | |
Solubility in water: 40g/L (20°C). Other solubilities: soluble in methanol, ethanol | |
Authentic | |
sodium;3-[[4-[(Z)-[4-(4-ethoxyanilino)phenyl]-[4-[ethyl-[(3-sulfonatophenyl)methyl]azaniumylidene]-2-methylcyclohexa-2,5-dien-1-ylidene]methyl]-N-ethyl-3-methylanilino]methyl]benzenesulfonate | |
6328534 | |
854.02 | |
Pure | |
Brilliant blue G |
Safety and Handling
Product Identifier
- Brilliant Blue G-250
RUO – Research Use Only