missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Brilliant Blue R, pure

Product Code. p-72009 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
5g
This item is not returnable. View return policy

Product Code. 10124260

Brand: Thermo Scientific Chemicals 191490050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6104-59-2 | C45H44N3NaO7S2 | 825.97 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6104-59-2
Molecular Formula C45H44N3NaO7S2
Molecular Weight (g/mol) 825.97
MDL Number MFCD00041762
InChI Key HAEGGLCXLFWTBE-UHFFFAOYSA-M
Synonym Acid Blue 83, C.I. 42660, Coomassie™ Brilliant Blue R-250
PubChem CID 61365
IUPAC Name sodium;3-[[4-[[4-(4-ethoxyanilino)phenyl]-[4-[ethyl-[(3-sulfonatophenyl)methyl]azaniumylidene]cyclohexa-2,5-dien-1-ylidene]methyl]-N-ethylanilino]methyl]benzenesulfonate
SMILES [Na+].CCOC1=CC=C(NC2=CC=C(C=C2)[C+](C2=CC=C(C=C2)N(CC)CC2=CC=CC(=C2)S([O-])(=O)=O)C2=CC=C(C=C2)N(CC)CC2=CC=CC(=C2)S([O-])(=O)=O)C=C1

Specifications

Absorbance 700 (at lambda min.)
Melting Point 174.0°C to 180.0°C
Absorption Ratio 1 to 1.2
Solubility Information Solubility in water: slightly soluble. Other solubilities: practically insoluble in ethanol
Infrared Spectrum Authentic
Lambda Max 588 to 596nm (in 50:50 ethanol:water)
Formula Weight 825.98
Grade Pure
Packaging Glass bottle
Color Bordeaux to Brown/Purple
Quantity 5 g
Physical Form Crystalline Powder
Chemical Name or Material Brilliant Blue R, Pure
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.