missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Brilliant Green Agar

Product Code. 12992588
Click to view available options
Quantity:
10/Pk.
Format:
90mm Monoplate
Media Type:
Brilliant Green Agar (Mod.)
Fill Weight:
19.5g ± 1.0g
Unit Size:
Pack of 10
This item is not returnable. View return policy

Product Code. 12992588

Brand: Thermo Scientific™ PO0171A

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Select and identify salmonellae (other than Salmonella typhi) fromfood, feed, water, and clinical samples with this diagnostic prepared media.

Select and identify salmonellae (other than Salmonella typhi) from food, feed, water, and clinical samples with Thermo Scientific™ Brilliant Green Agar (Modified) diagnostic medium. The modified formula provides improved inhibition of Escherichia coli and Proteus species. The medium has been widely assessed in Europe and has been used in ISO standards.

Brilliant Green Agar (Modified) provides greater inhibition of Escherichia coli and Proteus species than many other media formulations for the isolation of salmonellae. Growth of Pseudomonas species, whose colonies may resemble salmonellae on standard Brilliant Green Agar, are restricted, reducing confusion or extra work to confirm their identity. Inhibition of small numbers of salmonellae is absent from the modified formula1.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.

TRUSTED_SUSTAINABILITY

Specifications

Color Green / Brown
Content And Storage 2°C to 10°C
Format 90mm Monoplate
Media Type Brilliant Green Agar (Mod.)
Product Type Agar
Fill Weight 19.5g ± 1.0g
Packaging Type Wrapped in single cellulose-based film wrap
Quantity 10/Pk.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.