Learn More
Brilliant Stain Buffer Plus (RUO), BD Biosciences™
Buffer formulated for the immunofluorescent staining of cells.
Brand:  BD Biosciences 566385
Additional Details : Weight : 0.01000kg
Product Code. 15815009
  • £1580.00 / Each
Estimated Shipment: 29-07-2024
to see stock.
Description
Description
  • The BD Horizon Brilliant Stain Buffer Plus is a buffer formulated for the immunofluorescent staining of cells.
  • This buffer was developed to have a reduced test volume for applications where total staining volume is a concern
  • . Brilliant Stain Buffer Plus is as effective as BD Horizon Brilliant Stain Buffer (Cat. No. 563794/566349) in mitigating staining artifacts seen when using multiple BD Horizon Brilliant reagents.
  • For optimal and reproducible results, either BD Horizon Brilliant Stain Buffer Plus or BD Horizon Brilliant Stain Buffer should be used anytime two or more BD Horizon Brilliant™ dye-conjugated reagents are used in the same experiment.
  • The BD Horizon Brilliant Stain Buffer Plus is compatible with the use of other fluorescent staining reagents conjugated with traditional fluorochromes, such as fluorescein, phycoerythrin or Alexa Fluor™ dyes.
Specifications
Specifications
Brilliant stain buffer
1000 Tests