missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brivudine, 98%
Click to view available options
Quantity:
50 mg
250 mg
1 g
Unit Size:
1g
250mg
50mg
Beschreibung
It is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used as pharmaceutical intermediate.
Solubility
Soluble in water and methanol.
Notes
Store in cool dry conditions. Incompatible with oxidizing agents.
It is used as pharmaceutical intermediate.
Solubility
Soluble in water and methanol.
Notes
Store in cool dry conditions. Incompatible with oxidizing agents.
Spezifikation
Spezifikation
| Melting Point | ∽165°C (decomposition) |
| Quantity | 50 mg |
| Solubility Information | Soluble in water and methanol. |
| Optical Rotation | +10° (c=1 in Methanol) |
| Formula Weight | 333.14 |
| Percent Purity | 98% |
| Chemical Name or Material | (E)-5-(2-Bromovinyl)-2'-deoxyuridine |
RUO – Research Use Only
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur