missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brix Standard, Reagecon™
![BIOPHARMA](/content/dam/fishersci/glyphs/sure_trace.png)
Click to view available options
Concentration:
10% sucrose
30% sucrose
5% sucrose
60% sucrose
Description
This range of sucrose standard solutions contains high purity raw ingredients, can be used with any brand of refractometer and is stabilized to have an extended shelf-life to provide excellent value for users that are not required to follow ICUMSA Guidelines.
- Prepared gravimetrically on a weight/weight basis: both solute (sucrose) and solvent (water) are weighed on a balance calibrated using OIML traceable weights
- Balance Certificate of Calibration is issued in accordance with ISO 17025 for calibration of nonautomatic weighing machines (INAB ref: 265C)
- Brix value is verified using a high-performance calibrated, temperature-controlled refractometer using high-purity, ISO Guide 34-accredited Brix standards
Specifications
Description | Accredited to ISO 17025, In accordance with ISO Guide 34 |
Concentration | 10% sucrose |
Packaging | Dropper bottle |
Quantity | 15 mL |
For Use With (Equipment) | Refractometer |
IUPAC Name | (2R,3R,4S,5S,6R)-2-[(2S,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol |
Formula Weight | 342.3g/mol |
Solvent or Matrix | Water |
Solubility | Soluble in water |
Physical Form | Liquid |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'