missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRL 44408 maleate, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
BRL 44408 maleate is a selective α2A-adrenoceptor antagonist (Ki = 1.7 nM and 144.5 nM at α2A and α2B-adrenergic receptors respectively). Increases hippocampal noradrenalin release following systemic administration.
Specifications
Specifications
Quantity | 50 mg |
Formula Weight | 331.37 |
Percent Purity | >98% |
Chemical Name or Material | BRL 44408 maleate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'