Learn More
Hach Pacific Pty Ltd Bromcresol Green-Methyl Red Indicator Powder Pillows
Product Code. 15679524
Click to view available options
Quantity:
100/Pk.
Unit Size:
Pack of 100
Product Code. 15679524
Brand: Hach Pacific Pty Ltd 94399
pH indicator for determination of total alkalinity by titration methods.
pH indicator for determination of total alkalinity by titration methods. Pack of 100 Powder Pillows. pH indicator for determination of total alkalinity by titration methods. Pack of 100 Powder Pillows.
Specifications
Method: 8221
Method Name: Titration
Method Number 2: 8203
Number of tests: 100
Parameter: Alkalinity
Platform: Powder Pillows
Quantity: 100 /pk
Specifications
Quantity 100/Pk.
Type Water Testing Kit Supplies and Reagents