missing translation for 'onlineSavingsMsg'
Learn More
Bromfenac sodium, Thermo Scientific™ Product Code.: 17535565

Bromfenac sodium, Thermo Scientific™

Product Code. 17535565
250 mg
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy

Product Code. 17535565

Brand: Thermo Scientific Acros 468922500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 91714-93-1
Molecular Formula C15H11BrNNaO3
Molecular Weight (g/mol) 356.15
MDL Number MFCD03701673
InChI Key HZFGMQJYAFHESD-UHFFFAOYSA-M
IUPAC Name sodium 2-[2-amino-3-(4-bromobenzoyl)phenyl]acetate
SMILES [Na+].NC1=C(CC([O-])=O)C=CC=C1C(=O)C1=CC=C(Br)C=C1

Specifications

Packaging Glass Bottle
Quantity 250 mg
Certificate of Analysis Available
Physical Form Powder
Color Yellow to Orange
Chemical Name or Material Bromfenac sodium
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Bromfenac sodium, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.