missing translation for 'onlineSavingsMsg'
Learn More

Bromide QC Sample, Wastewater Standard, SPEX CertiPrep™

Product Code. 15698215 Shop All SPEX Certiprep Products
Click to view available options
Quantity:
2 mL
Unit Size:
2mL
This item is not returnable. View return policy

Product Code. 15698215

Brand: SPEX Certiprep 769

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

Bromide Water Pollution QC Sample for use with ion chromatography or colorimetric methods, 15 mL, SPEX CertiPrep™

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

  • Bromide QC Sample
  • QC Sample for wastewater analysis
  • 15 mL volume

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Packaging Triple-leached LDPE bottle
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Type Certified Reference Material
Industry Type Wastewater Testing
Quantity 2 mL
Chemical Name or Material Bromide QC Sample, Yields up to 1L after dilution

Product intended for laboratory use only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.

Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt