missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bromine, 1M solution in acetic acid, AcroSeal™
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle
Unit Size:
100mL
800mL
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent
Specifications
Specifications
| Chemical Name or Material | Bromine |
| Color | Red |
| Name Note | 1M solution in acetic acid |
| CAS | 7726-95-6, 64-19-7 |
| Molecular Formula | Br |
| Linear Formula | Br2 |
| MDL Number | MFCD00010896 |
| Concentration | 0.90 to 1.10M |
| Density | 3.111 |
| Merck Index | 15, 1401 |
| Show More |
Safety and Handling
Product Identifier
- Bromine, 1M solution in acetic acid
- Danger
- Acute toxicity Category 1
- ACUTE AQUATIC Acute 1
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 3
- Skin corrosion/irritation Category 1 A
- H318-Causes serious eye damage.
- H330-Fatal if inhaled.
- H400-Very toxic to aquatic life.
- H314-Causes severe skin burns and eye damage.
- H226-Flammable liquid and vapour.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction