missing translation for 'onlineSavingsMsg'
Learn More

Bromine, 99.5+%, Certified AR for Analysis, Liquid, Fisher Chemical™

£87.74 - £340.58

Chemical Identifiers

CAS 7726-95-6
Molecular Formula Br
Molecular Weight (g/mol) 79.91
MDL Number 10896
InChI Key CPELXLSAUQHCOX-UHFFFAOYSA-M
IUPAC Name bromide
SMILES [Br-]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11463533
SDS View Documents
Fisher Chemical
B/4050/PB05
100 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11473533
SDS View Documents
Fisher Chemical
B/4050/PB08
500 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Specifications
Show More
Videos
Safety and Handling
Show More
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Bromine, 99.5+%, Certified AR for Analysis, Liquid, Fisher Chemical™

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.