missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bromine, 99+%, extra pure, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 mL
500 mL
1 L
Packaging:
Plastic Coated Glass Bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Color | Red to Brown |
Physical Form | Fuming Liquid |
Quantity | 1 L |
Assay Percent Range | 99+% |
Linear Formula | Br2 |
Fieser | 03,34; 04,46; 05,55; 06,70; 07,33; 08,52; 09,65; 10,56; 11,75; 12,70; 13,47; 14,56; 15,47 |
Merck Index | 15, 1401 |
IUPAC Name | molecular bromine |
Formula Weight | 159.82 |
Percent Purity | ≥99.0% |
Show More |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0468737 | Certificate of Analysis | 31/01/2025 | 402845000,402841000,402840010 |
A0468997 | Certificate of Analysis | 31/01/2025 | 402845000,402841000,402840010 |
A0461949 | Certificate of Analysis | 11/04/2024 | 402845000,402840010,402841000 |
A0453635 | Certificate of Analysis | 25/08/2023 | 402845000,402840010,402841000 |
A0451155 | Certificate of Analysis | 02/05/2023 | 402845000,402840010,402841000 |
Safety and Handling



Product Identifier
- Bromine
- Danger
- Skin corrosion/irritation Category 1 A
- Serious eye damage/eye irritation Category 1
- Acute toxicity Category 1
- ACUTE AQUATIC Acute 1
- H314-Causes severe skin burns and eye damage.
- H330-Fatal if inhaled.
- H400-Very toxic to aquatic life.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P273-Avoid release to the environment.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Bromine, 99+%, extra pure, Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction