missing translation for 'onlineSavingsMsg'
Learn More

Bromine concentrate, For 1L standard solution, 0.05 M Br2, Honeywell Fluka™

Product Code. p-7122093 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
1 Pc.
6 x 1 Ea.
Unit Size:
Each
Pack of 6
This item is not returnable. View return policy

Product Code. 15655190

Brand: Honeywell-Fluka 380401EA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

for 1L standard solution, 0.05 M Br2

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7726-95-6
Molecular Formula Br
Molecular Weight (g/mol) 79.91
InChI Key CPELXLSAUQHCOX-UHFFFAOYSA-M
Synonym bromine, dibromine, brom, bromine solution, brome, bromo, broom, bromine water, bromo italian, bromo spanish
PubChem CID 24408
ChEBI CHEBI:29224
IUPAC Name molecular bromine
SMILES [Br-]

Specifications

Quantity 1 Pc.
MDL Number MFCD00010896
UN Number NONH for all modes of transport
Formula Weight 159.81g/mol
Density 1.307g/mL at 25°C
Chemical Name or Material Bromine concentrate
compliance-icons
Product Identifier
  • Bromine concentrate
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 1B
Hazard Statement
  • H350-May cause cancer.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : potassium bromate, Potassium bromide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.