missing translation for 'onlineSavingsMsg'
Learn More

Bromine, Extra Pure, SLR, Fisher Chemical™

Product Code. 10182180 Shop All Fisher Chemical Products
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
This item is not returnable. View return policy

Product Code. 10182180

Brand: Fisher Chemical B/4000/PB08

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 7726-95-6
Molecular Formula Br
Molecular Weight (g/mol) 79.91
MDL Number 10896
InChI Key CPELXLSAUQHCOX-UHFFFAOYSA-M
IUPAC Name bromide
SMILES [Br-]

Specifications

Boiling Point 58.7°C
Melting Point -7.2°C
Color Brown-Red
Physical Form Liquid
Quantity 500 mL
Formula Weight 159.82g/mol
Packaging Plastic Coated Glass bottle
Vapor Pressure 230mbar at 20°C
Density Vapor Density: 5.51
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Bromine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H318-Causes serious eye damage.
  • H400-Very toxic to aquatic life.
  • H314-Causes severe skin burns and eye damage.
  • H330-Fatal if inhaled.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.