missing translation for 'onlineSavingsMsg'
Learn More

Bromine, Extra Pure, SLR, Fisher Chemical™

Product Code. p-7072868 Shop All Fisher Chemical Products
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
This item is not returnable. View return policy

Product Code. 10510721

Brand: Fisher Chemical B/4000/PB05

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Currently Unavailable: We have no information when this product will be available.
View alternative products
This item is not returnable. View return policy

Chemical Identifiers

CAS 7726-95-6
Molecular Formula Br
Molecular Weight (g/mol) 79.91
MDL Number 10896
InChI Key CPELXLSAUQHCOX-UHFFFAOYSA-M
IUPAC Name bromide
SMILES [Br-]

Specifications

Boiling Point 58.7°C
Melting Point -7.2°C
Color Brown-Red
Physical Form Liquid
Quantity 100 mL
Formula Weight 159.82g/mol
Packaging Plastic Coated Glass bottle
Vapor Pressure 230mbar at 20°C
Density Vapor Density: 5.51
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.